Learn More
Smiths Medical Hypodermic Needle-Pro™ Device and Syringe
Rotating safety device facilitates intradermal injections and does not contribute to additional dead space to standard needle/syringe injections
Supplier: Smiths Medical LSEAL.CORE
This item is not returnable.
View return policy
Description
- Meets NIOSH/CDC recommendations for safety devices
- Safety feature engages with a single-handed technique
- Facilitates two-needle injection techniques