Smiths Medical Pro-Vent™ Plus Arterial Blood Gas Sampling 1cc/7IU Kits
Proven safety devices, quick-filling syringe, advanced formula heparin
Manufacturer: Smiths Medical PM, Inc. 4640LH
 View more versions of this product
Catalog No. 22-024-945
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    For Use With (Application) Allows healthcare professionals to obtain very small blood samples and still maintain the full range of blood gas, electrolyte and metabolite testing with reduced heparin interference and extended clotting time
    Includes 1mL luer-slip syringe, Filter-Pro™ device, 25G x 0.62 in. preattached needle with Needle-Pro™ needle safety device 3G x 1 in. secondary needle with Needle-Pro™ device attached, support kit, and iodophor prep pad
    Packaging Sterile Tyvek™
    Shelf Life 3 Years
    Sterile Yes
    Capacity (Metric) 1mL
    Needle Gauge 23 to 25 ga
    Product Type Arterial Blood Sampling Kit
    Quantity 100 Case
    Volume (Metric) 1mL
  • Description & Specifications
Related Products
Product Content Feedback