Smiths Medical Pro-Vent™ Plus Arterial Blood Gas Sampling 3cc/70.5IU Kits

Proven safety devices, quick-filling syringe, advanced formula heparin

Manufacturer: Smiths Medical PM, Inc. 4699P1

 View more versions of this product

Catalog No. 22-004-178

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    For Use With (Application) Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results
    Includes 3mL luer-slip syringe with Filter-Pro™ device, 23G x 1 in. preattached needle with Needle-Pro™ needle safety device 22G x 1.5 in. secondary needle, support kit
    Packaging Sterile Tyvek™
    Shelf Life 3 Years
    Sterile Yes
    Capacity (Metric) 3mL
    Needle Gauge 22 to 23 ga
    Product Type Arterial Blood Sampling Kit
    Quantity 100 Case
    Volume (Metric) 3mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit