Learn More
ICU Medical Jelco™ Saf-T Holder™ Blood Tube Holders
Saf-T Holder™ devices come with the back-end needle and multi-sample Luer permanently attached to the holder to minimize potential exposures and holder re-use. Available separately or with Saf-T Wing™ Blood Collection & Infusion Sets, Saf-T Holder™ devices also provide easy needless transfer after a syringe draw.
Supplier:  ICU Medical 96007
 View more versions of this product
Catalog No. 22-024-902
Add to Cart
Description
Description
For use with Saf-T Wing™ winged infusion sets and Saf-T Closed Blood Collection System™ sets.
  • Back-end needle and multi-sample Luer permanently attached to the holder to eliminate potential exposures and holder re-use
  • Available with male or female Luer adapter
  • Sterilized with ethylene oxide
  • Latex-free
Order Info
Standard length holder fits most blood collection systems. Shorter length designed for use exclusively with Trek Diagnostic systems.
Certifications
CE marked
Specifications
Specifications
Non-sterile
Yes
CE, OSHA
Provide safe needleless transfer after a syringe draw
Saf-T Wing winged infusion sets and Saf-T closed Blood collection system
Pediatric Tube Insert
Pediatric insert
Safety and Handling
Safety and Handling
ShelfLife : 5 Years
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'