Learn More
SMN Mouse anti-Human, Mouse, Rat, Primate, Xenopus, Zebrafish, Alexa Fluor™ 350, Clone: 2B1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB1001936AF350
Catalog No. N101936A350
Add to cart
Description
Description
SMN Monoclonal antibody specifically detects SMN in Human,Mouse,Rat,Primate,Xenopus,Zebrafish samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
SMN
2B1
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gemin-1, Kugelberg-Welander disease), SMNC, survival motor neuron protein, survival of motor neuron 1, telomeric
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Primate, Rat, Xenopus, Zebrafish
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
SMN1
Purified recombinant His6-tagged human SMN protein. [Swiss-ProtNo. Q16637]
0.1 mL
Primary
6606