Thermo Scientific™ Smoke-Check Smoke Detector Challenge Particles Available on GSA/VA Contract for Federal Government customers only.
Check transport time of air sampling smoke detectors in cleanrooms and other critical environments.
Manufacturer: Thermo Scientific™ SD01
 View more versions of this product
Catalog No. 09-980-571
  Check Availability
Add to Cart
    Specifications
    Type Specialty Particles
    For Use With (Application) Air Sampling Transport Time
    Description Smoke Detector Challenge Particles
    Quantity 10mL
    Check transport time of air sampling smoke detectors as required by United States NFPA Standard 72 with Thermo Scientific™ Smoke-Check Smoke Detector Challenge Particles, especially useful in cleanrooms and other critical environments.
    A suspension of polystyrene particles in high purity water is nebulized using a handheld ultrasonic atomizer to mimic natural smoke, thereby eliminating residues caused by traditional oil-based sprays or real smoke.
    • Particle size and concentration have been optimized to consistently trigger the smoke detector
    • Will not pass through HEPA filters, ensuring the integrity of the cleanroom environment
    Smoke Detector Challenge Kit:
    Includes a battery powered portable ultrasonic nebulizer, a draft shield and a 10mL bottle of particles.
    Ordering Information:
    Additional bottles of Smoke-Check may be purchased separately.
  • Description & Specifications
Related Products
Product Content Feedback