Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guardian™ IN-35, Smooth Finish Neoprene Gloves
SDP

Smooth finish 35 mil neoprene gloves provide protection from petroleum, oils and lubricants (POLs), including fueling operations.

Manufacturer:  Guardian™ 49910

 View more versions of this product

Catalog No. 19-321-136


Add to cart
This item is not returnable. View return policy

Description

Description

  • Fitted hand style with smooth finish texture
  • Protects against wide range of hazardous chemicals
  • Flame resistant and vapor protective
  • Meets DOD# MIL-DTL-12223 standards
  • Reusable and easy cleaning
  • 35-mil thickness
  • 14 in. length
Specifications

Specifications

ASTM D412, ASTM F739
Black
Yes
10
0.88 mm
Smooth
ISO 9001:2000
IN-35
35.5 cm
CE
Specific
Neoprene
35 mil
Beaded/Rolled
Unlined
49910
14 in.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit