Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Guardian™ IN-35, Smooth Finish Neoprene Gloves

Click to view available options
Size:
8
9
10
11
12
Description
- Fitted hand style with smooth finish texture
- Protects against wide range of hazardous chemicals
- Flame resistant and vapor protective
- Meets DOD# MIL-DTL-12223 standards
- Reusable and easy cleaning
- 35-mil thickness
- 14 in. length
Specifications
Certifications/Compliance | ASTM D412, ASTM F739 |
Certifications/Compliance (Europe) | CE |
Color | Black |
Hand | Specific |
Latex Free | Yes |
Material | Neoprene |
Size | 11 |
Thickness (English) | 35 mil |
Thickness (Metric) | 0.88 mm |
Glove Cuff Style | Beaded/Rolled |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title