Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guardian™ IN-35, Smooth Finish Neoprene Gloves
SDP

Catalog Number 19321138
Click to view available options
Size:
8
9
10
11
12
This item is not returnable. View return policy
This item is not returnable. View return policy

Smooth finish 35 mil neoprene gloves provide protection from petroleum, oils and lubricants (POLs), including fueling operations.

  • Fitted hand style with smooth finish texture
  • Protects against wide range of hazardous chemicals
  • Flame resistant and vapor protective
  • Meets DOD# MIL-DTL-12223 standards
  • Reusable and easy cleaning
  • 35-mil thickness
  • 14 in. length
Certifications/Compliance ASTM D412, ASTM F739
Certifications/Compliance (Europe) CE
Color Black
Hand Specific
Latex Free Yes
Material Neoprene
Size 12
Thickness (English) 35 mil
Thickness (Metric) 0.88 mm
Glove Cuff Style Beaded/Rolled
Grip Design Smooth
Lining Unlined
Manufacturing/QMS Audit Standards ISO 9001:2000
Model 49912
Series IN-35
Length (English) 14 in.
Length (Metric) 35.5 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit