Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guardian™ IN-35, Smooth Finish Neoprene Gloves
SDP

Smooth finish 35 mil neoprene gloves provide protection from petroleum, oils and lubricants (POLs), including fueling operations.

$68.00

Specifications

Certifications/Compliance ASTM D412, ASTM F739
Certifications/Compliance (Europe) CE
Manufacturing/QMS Audit Standards ISO 9001:2000
Color Black
Glove Cuff Style Beaded/Rolled
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-321-134
SDP
View Documents
Guardian™
49908
8
Pair for $68.00
Add to cart
 
19-321-135
SDP
View Documents
Guardian™
49909
9
Pair for $68.00
Add to cart
 
19-321-136
SDP
View Documents
Guardian™
49910
10
Pair for $68.00
Add to cart
 
19-321-137
SDP
View Documents
Guardian™
49911
11
Pair for $68.00
Add to cart
 
19-321-138
SDP
View Documents
Guardian™
49912
12
Pair for $68.00
Add to cart
 
Description

Description

  • Fitted hand style with smooth finish texture
  • Protects against wide range of hazardous chemicals
  • Flame resistant and vapor protective
  • Meets DOD# MIL-DTL-12223 standards
  • Reusable and easy cleaning
  • 35-mil thickness
  • 14 in. length
Specifications

Specifications

ASTM D412, ASTM F739
ISO 9001:2000
Beaded/Rolled
14 in.
35 mil
CE
Black
Smooth
35.5 cm
0.88 mm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit