Learn More
Alfa Aesar™ Smoothened Agonist Dihydrochloride Dihydrate
A highly potent activator of Smoothened
Manufacturer:  Alfa Aesar™ J64726LB0
 View more versions of this product
Catalog No. AAJ64726LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
5mg
2°C to 8°C
Dihydrochloride Dihydrate
364590-63-6
599.00
Light Sensitive and Hygroscopic
Pale yellow
Solid
A highly potent activator of Smoothened
C28H28ClN3OS.2HCl.2H2O
SAG1.3 HCl