Learn More
Smoothened Mouse anti-Human, Clone: 1015418, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems MAB10276100
 View more versions of this product
Catalog No. MAB10276100
Add to cart
This item is not returnable. View return policy
Description
Description
Smoothened Monoclonal antibody specifically detects Smoothened in Human samples. It is validated for Immunohistochemistry.
Specifications
Specifications
Smoothened
1015418
Immunohistochemistry 1-25 ug/mL
Q99835
bnb, CRJS, FZD11, Gx, Protein Gx, SMO, SMOH, SMOHseven transmembrane helix receptor, smoothened (Drosophila) homolog, smoothened homolog, smoothened homolog (Drosophila)
Chinese Hamster Ovary cell line, CHO-derived human Smoothened, Met1-Tyr233, Accession # Q99835
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
6608
Reconstitute at 0.5 mg/mL in sterile PBS.
Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Purified
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Monoclonal
Detects human Smoothened in direct ELISAs.
Human