Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Smoothened Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB10276SP
This item is not returnable.
View return policy
Description
Smoothened Monoclonal specifically detects Smoothened in Human samples. It is validated for Immunohistochemistry.Specifications
Smoothened | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
bnb, CRJS, FZD11, Gx, Protein Gx, SMO, SMOH, SMOHseven transmembrane helix receptor, smoothened (Drosophila) homolog, smoothened homolog, smoothened homolog (Drosophila) | |
Chinese Hamster Ovary cell line, CHO-derived human Smoothened, Met1-Tyr233, Accession # Q99835 | |
25 μg | |
Primary | |
Detects human Smoothened in direct ELISAs. | |
Human | |
Purified |
Immunohistochemistry | |
1015418 | |
Immunohistochemistry 1-25 ug/mL | |
Q99835 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6608 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction