Smopex-101, Alfa Aesar™

For Recovery of Precious Metals From Catalyst Process Streams, Especially Pd, Rh

Manufacturer: Alfa Aesar 4456814

Quantity 25g

 View more versions of this product

Catalog No. AA4456814

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Fibers
    MDL Number MFCD06202258
    Synonym Styrene sulfonic acid grafted polyolefin fiber
    Chemical Name or Material Smopex-101

    Specifications

    Physical Form Fibers
    Quantity 25g
    Color Yellow
    Density 0.3g/cm3 at 20°C
    MDL Number MFCD06202258
    Synonym Styrene sulfonic acid grafted polyolefin fiber
    Odor Odorless
    Chemical Name or Material Smopex-101

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit