Alfa Aesar™ Smopex™-102, metal scavenger

Manufacturer: Alfa Aesar™ 4470514

Quantity 25g

 View more versions of this product

Catalog No. AA4470514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 289911-93-9
    MDL Number MFCD06202259
    Synonym Acrylic acid grafted polyolefin fiber
    Chemical Name or Material Smopex™-102 metal scavenger

    Specifications

    Density 0.3g/mL
    MDL Number MFCD06202259
    CAS 289911-93-9
    Synonym Acrylic acid grafted polyolefin fiber
    Physical Form Powder
    Color White to pale brown
    Quantity 25g
    Chemical Name or Material Smopex™-102 metal scavenger

    Description

    • Recovery of precious metals from catalyst process streams, especially Pt
    • Also, base metals removed: Fe, Cu, and Ni

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit