Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Smopex™-102, metal scavenger

Manufacturer:  Alfa Aesar™ 4470522

Quantity 100g

 View more versions of this product

Catalog No. AA4470522


  Check Availability
Add to cart

Specifications

Description

  • Recovery of precious metals from catalyst process streams, especially Pt
  • Also, base metals removed: Fe, Cu, and Ni
Description & Specifications

Chemical Identifiers

289911-93-9
MFCD06202259
Acrylic acid grafted polyolefin fiber
Smopex™-102 metal scavenger

Specifications

289911-93-9
Powder
Acrylic acid grafted polyolefin fiber
100g
0.3g/mL
MFCD06202259
White to pale brown
Smopex™-102 metal scavenger
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit