Alfa Aesar™ Smopex™-103, metal scavenger

Manufacturer: Alfa Aesar™ 4470614

Quantity 25g

 View more versions of this product

Catalog No. AA4470614

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 527752-26-7
    MDL Number MFCD06202260
    Synonym Styryl dimethylamine grafted polyolefin fiber
    Chemical Name or Material Smopex™-103 metal scavenger

    Specifications

    MDL Number MFCD06202260
    CAS 527752-26-7
    Synonym Styryl dimethylamine grafted polyolefin fiber
    Color White
    Physical Form Powder
    Quantity 25g
    Chemical Name or Material Smopex™-103 metal scavenger

    Description

    • Recovery of precious metals from catalyst process streams, specifically Os
    • Also, heavy metals removed : Re, Au, Co, and Cr

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit