Learn More
Alfa Aesar™ Smopex™-103, metal scavenger
Manufacturer:  Alfa Aesar™ 4470622
Quantity 100g
 View more versions of this product
Catalog No. AA4470622
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • Recovery of precious metals from catalyst process streams, specifically Os
  • Also, heavy metals removed : Re, Au, Co, and Cr
Description & Specifications
Chemical Identifiers
527752-26-7
MFCD06202260
Styryl dimethylamine grafted polyolefin fiber
Smopex™-103 metal scavenger
Specifications
527752-26-7
MFCD06202260
White
Smopex™-103 metal scavenger
Powder
Styryl dimethylamine grafted polyolefin fiber
100g