Smopex™-105, Alfa Aesar™
For Recovery of Negatively Charged Precious Metals Complexes Used in Complex Reactions (Suzuki, Heck, Sonagashira). Recovery of Precious Metals From Catalyst Process Streams
Manufacturer: Alfa Aesar 4471022
Quantity 100g
 View more versions of this product
Catalog No. AA4471022
  Check Availability
Add to Cart
Related Products
Product Content Feedback