Learn More
Smopex™-105, Alfa Aesar™
For Recovery of Negatively Charged Precious Metals Complexes Used in Complex Reactions (Suzuki, Heck, Sonagashira). Recovery of Precious Metals From Catalyst Process Streams
Manufacturer:  Alfa Aesar 4471006
Quantity 5g
 View more versions of this product
Catalog No. AA4471006
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
Fibers
MFCD06202261
Vinyl pyridine grafted polyolefin fiber
Smopex-105, Weak base (pyridine functional group) anion exchanger. Exchange capacity 4-6mmol/g
Specifications
Fibers
Vinyl pyridine grafted polyolefin fiber
White
Smopex-105, Weak base (pyridine functional group) anion exchanger. Exchange capacity 4-6mmol/g
MFCD06202261
Odorless
5g