Alfa Aesar™ Smopex™-105 FB, metal scavenger

Manufacturer: Alfa Aesar™ 4727222

Quantity 100g

 View more versions of this product

Catalog No. AA4727222

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Synonym Vinyl pyridine grafted polyolefin fiber (free base)
    Chemical Name or Material Smopex™-105 FB, Metal scavenger

    Specifications

    Synonym Vinyl pyridine grafted polyolefin fiber (free base)
    Color White to biege to yellow
    Quantity 100g
    Physical Form Fibers
    Odor Odorless
    Chemical Name or Material Smopex™-105 FB, Metal scavenger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit