Alfa Aesar™ Smopex™-110, metal scavenger

In precious metal recovery from Suzuki, Heck, Sonagashira and Shille type reactions

Manufacturer: Alfa Aesar™ 4472409

Quantity 10g

 View more versions of this product

Catalog No. AA4472409

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD06202263
    Chemical Name or Material Smopex™-110 metal scavenger

    Specifications

    Physical Form Powder
    MDL Number MFCD06202263
    Quantity 10g
    TSCA TSCA
    Chemical Name or Material Smopex™-110 metal scavenger

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit