Learn More
Alfa Aesar™ Smopex™-110, metal scavenger
In precious metal recovery from Suzuki, Heck, Sonagashira and Shille type reactions
Manufacturer:  Alfa Aesar™ 4472404
Quantity 2g
 View more versions of this product
Catalog No. AA4472404
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Chemical Identifiers
MFCD06202263
Smopex™-110 metal scavenger
Specifications
Powder
MFCD06202263
2g
Smopex™-110 metal scavenger
Safety and Handling
TSCA : TSCA