Alfa Aesar™ Smopex™-112v, metal scavenger

Manufacturer: Alfa Aesar™ 4547414

Quantity 25g

 View more versions of this product

Catalog No. AA4547414

  Check Availability
Add to Cart

    Chemical Identifiers

    Synonym Acrylate based 'alpha'-hydroxyl thiol grafted polyolefin fiber
    Chemical Name or Material Smopex™-112v metal scavenger

    Specifications

    Physical Form Powder
    Synonym Acrylate based 'alpha'-hydroxyl thiol grafted polyolefin fiber
    Color Off-white
    Quantity 25g
    Chemical Name or Material Smopex™-112v metal scavenger

    Description

    • Recovery of precious metals from catalyst processes, especially Pd and Pt
    • Also, base metals removed: Ag
    • This product is especially suited for aqueous applications

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit