Learn More
Alfa Aesar™ Smopex™-234, metal scavenger
Manufacturer:  Alfa Aesar™ 4498414
Quantity 25g
 View more versions of this product
Catalog No. AA4498414
  Check Availability
Add to cart
Specifications
Description
  • Recovery of precious metals from catalyst processes, especially Pd, Rh, Ru, Pt, and Ir
  • Also, base metals removed: Sn
Description & Specifications
Chemical Identifiers
912338-63-7
Mercaptoethylacrylate grafted polyolefin fiber
Smopex™-234 metal scavenger
Specifications
912338-63-7
Mercaptoethylacrylate grafted polyolefin fiber
25g
Powder
Off-white
Smopex™-234 metal scavenger