Alfa Aesar™ Smopex™-301, metal scavenger

Recovery of precious metals from catalyst processes, especially Pd, Ru, and Ir

Manufacturer: Alfa Aesar™ 4590222

Quantity 100g

 View more versions of this product

Catalog No. AA4590222

  Check Availability
Add to Cart

    Chemical Identifiers

    Synonym Styryl diphenylphosphine grafted polyolefin fiber
    Chemical Name or Material Smopex™-301 metal scavenger

    Specifications

    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Fibers
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H361
    Suspected of damaging fertility or the unborn child.
    Synonym Styryl diphenylphosphine grafted polyolefin fiber
    Color Off-white
    Quantity 100g
    Chemical Name or Material Smopex™-301 metal scavenger

    GHS H Statement
    H361
    Suspected of damaging fertility or the unborn child.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit