Learn More
Alfa Aesar™ Smopex™-301, metal scavenger
Recovery of precious metals from catalyst processes, especially Pd, Ru, and Ir
Manufacturer:  Alfa Aesar™ 4590206
Quantity 5g
 View more versions of this product
Catalog No. AA4590206
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
Styryl diphenylphosphine grafted polyolefin fiber
Smopex™-301 metal scavenger
Specifications
Fibers
Off-white
Smopex™-301 metal scavenger
Styryl diphenylphosphine grafted polyolefin fiber
5g
Safety and Handling
SDS & Safety Documents
GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning