Learn More
LIFE TECHNOLOGIES Platinum™ Taq Green Hot Start DNA Polymerase 120rxn DFS Item
Manufacturer:  LIFE TECHNOLOGIES 11966010
Each box contains:• 24 µL Platinum Taq DNA Polymerase• 1.25 mL 10X Green PCR Buffer, -Mg• 1.25 mL 50 mM magnesium chloride• 1.25 mL KB ExtenderStore at -20°C in a non-frost-free freezer.
Catalog No. 11-966-010