Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMPDL3B 293T Cell Overexpression Lysate (Denatured), Abnova
Supplier: Abnova Corporation H00027293T01
Description
Western Blotting
Specifications
Western Blot | |
SMPDL3B 293T Cell Transient Overexpression Lysate(Denatured) | |
41.14 | |
pCMV-SMPDL3B full-length | |
100 μL | |
Human |
293T | |
NM_001009568.1 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-SMPDL3B antibody (H00027293-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction