Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMPX Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542030
Description
Peptide sequence: TPEVEEGVPP TSDEEKKPIP GAKKLPGPAV NLSEIQNIKS ELKYVPKAEQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 79%; Rabbit: 100%; Rat: 100%.
SMPX plays a role in the regulatory network through which muscle cells coordinate their structural and functional states during growth, adaptation, and repair.
Specifications
SMPX | |
Polyclonal | |
Unconjugated | |
SMPX | |
1010001C09Rik; cardiac and skeletal muscle protein; chisel protein; Csl; deafness, X-linked 6, sensorineural; DFN6; DFNX4; small muscle protein X-linked; small muscle protein, X-linked; small muscular protein; Smpx; Srmx; Stretch-responsive skeletal muscle protein; Unknown (protein for MGC:128317) | |
Rabbit | |
Protein A | |
RUO | |
23676 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UHP9 | |
SMPX | |
Synthetic peptide directed towards the middle region of human SMPX. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction