Learn More
SMU1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 155111AP
Catalog No. 50-173-0279
Add to cart
Description
Description
This antibody is a rabbit polyclonal antibody raised against a region of human REDD1 antigen.
SMU1 (suppressor of Mec8 and Unc-52 homolog), also known as BWD (brain-enriched WD), is a member of the WD-repeat SMU1 family and contains one CTLH domain, one LisH domain and six WD-repeats. SMU1 is a homolog of the C. elegans protein Smu1, a ubiquitously expressed nuclear protein that is believed to play a role in alternative splicing events of Unc-52. In mammals, SMU1 is a component of the spliceosome and appears to exhibit a conserved function, affecting the splicing of the mammalian Unc-52 homolog, namely Perlecan. Mutations in the gene encoding SMU1 may affect the function of the spliceosome.
Specifications
Specifications
SMU1
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q2TAY7, Q3UKJ7, Q99M63
BWD, fSAP57, RP11 54K16.3, SMU 1, SMU1
Rabbit
IgG
150 μL
-20°C
Primary
117541, 55234, 74255
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SMU1
Liquid
SMU1
SMU1 Fusion Protein Ag7688
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat