Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMVT/SLC5A6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen SMVT/SLC5A6
Applications Immunohistochemistry
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092890
View Documents
Novus Biologicals
NBP302997100UL
100 μg
Each for $320.71
Add to cart
 
NB092891
View Documents
Novus Biologicals
NBP30299720UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

SMVT/SLC5A6 Polyclonal antibody specifically detects SMVT/SLC5A6 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry
Specifications

Specifications

SMVT/SLC5A6
Unconjugated
Rabbit
Human, Mouse, Rat
Na(+)-dependent multivitamin transporter, SMVTNa+-dependent multivitamin transporter, sodium-dependent multivitamin transporter, solute carrier family 5 (sodium-dependent vitamin transporter), member 6, Solute carrier family 5 member 6
A synthetic peptide corresponding to a sequence within amino acids 500 to the C-terminus of human SMVT/SLC5A6 (NP_066918.2).
Polyclonal
Affinity purified
Immunohistochemistry
Purified
RUO
PBS with 50% glycerol, pH7.3.
8884
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit