Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMYD4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA551142
Description
The antibody detects endogenous levels of total SMYD4 protein.
SMYD4 Polyclonal antibody specifically detects SMYD4 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
SMYD4 | |
Polyclonal | |
Unconjugated | |
SMYD4 | |
G430029E23Rik; Kiaa1936; SET and MYND domain containing 4; SET and MYND domain-containing protein 4; Smyd4; ZMYND21 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114826 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8IYR2 | |
SMYD4 | |
Fusion protein of human SMYD4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction