Learn More
Echelon Biosciences Research LabsSupplier Diversity Partner Zn (II) Deuteroporphyrin IX 2,4 bisethylene glycol 25 mg  DFS Item
Manufacturer:  Echelon Biosciences Research LabsSupplier Diversity Partner FH09025MG
Zn (II) Deuteroporphyrin IX 2,4 bis ethyleneglycol is an inhibitor of heme oxygenase (the enzyme that catalyzes the conversion of heme to biliverdin in the heme degradation pathway) at concentrations as low at 0.5 μM. Heme oxygenase has been implicated in tumor cell resistance to chemotherapy, reduction of free radical formation and inflammation, and associated with vascular repair.
Catalog No. 50-720-336