Thermo Scientific™ Snap-Cap Centrifuge Tubes, Plain Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Plain Snap-Cap Centrifuge Tubes include a flat cap, and have no frosting or graduations.
Manufacturer: Thermo Scientific™ 3444
 View more versions of this product
Catalog No. 21-403-176
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Capacity (Metric) 1.5mL
    Sterility Nonsterile
    Max. RCF 25,000 x G
    Color Natural,Natural
    Material Polypropylene
    Sterile No,No
    Cap Style Flat
    Packaging 10 bags of 50
    Certifications/Compliance Certified free of RNase, DNase, DNA and pyrogens
    Description Plain, Snap-Cap Centrifuge Tube
    Capacity 1.5mL
    Thermo Scientific™ Plain Snap-Cap Centrifuge Tubes include a flat cap, and have no frosting or graduations.
    • With flat cap
    • No frosting or graduations
    • Certified RNase/DNase, RNA/DNA, ATP and pyrogen free
  • Description & Specifications
Related Products
Product Content Feedback