Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RayDyLyo™ Snap Caps from DWK Life Sciences™
SDP

These plastic push-fit caps are designed for closing vials of injectable drugs. Flip-off snap caps are pre-assembled with an ISO 8362-2 compliant elastomer stopper to simplify the production process.

Supplier:  RayDyLyo™ 255855001

 View more versions of this product

Catalog No. 01-000-877


Explore available promotions
Add to Cart
Request a Quote
This item is not returnable. View return policy

Description

Description

  • Container Closure Integrity (CCI) validated for use with ISO 8362-1 glass and COP vials
  • Delivered sterile (RTU) with a 2 year shelf life from time of manufacture
  • Clean room manufactured with 100% in-process quality controls
  • Suitable for aseptic small batch, clinical trial, and robotic fill and finish operations
  • Snap caps do not require special crimping tools and affix with single vertical pressure
  • Provides tamper-evident closing and eliminates crimping defects
  • Compatible with gamma and steam terminal sterilization
  • Available in nested, standard bag, and RTP-type bag configurations
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blue
Polypropylene
Bagged
3200/Pk.
Igloo Lyophilization Vials
13 mm
Sterile
Autoclavable
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

RayDyLyo™ is a registered trademark of ARaymond.