Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RayDyLyo™ Snap Caps from DWK Life Sciences™

Click to view available options
Closure Size:
13 mm
20 mm
Color:
Blue
Red
Packaging Type:
Bagged
Nested
For Use With (Equipment):
Igloo Lyophilization Vials
Serum Vials
Description
- Container Closure Integrity (CCI) validated for use with ISO 8362-1 glass and COP vials
- Delivered sterile (RTU) with a 2 year shelf life from time of manufacture
- Clean room manufactured with 100% in-process quality controls
- Suitable for aseptic small batch, clinical trial, and robotic fill and finish operations
- Snap caps do not require special crimping tools and affix with single vertical pressure
- Provides tamper-evident closing and eliminates crimping defects
- Compatible with gamma and steam terminal sterilization
- Available in nested, standard bag, and RTP-type bag configurations

Specifications
Specifications
Color | Blue |
Diameter (Metric) Outer | 13 mm |
Material | Polypropylene |
Sterility | Sterile |
Packaging Type | Nested |
Autoclavable | Autoclavable |
Quantity | 3600/Pk. |
Certifications/Compliance | ISO 8362-2 |
Closure Size | 13 mm |
For Use With (Equipment) | Igloo Lyophilization Vials |
RayDyLyo™ is a registered trademark of ARaymond.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction