Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RayDyLyo™ Snap Caps from DWK Life Sciences™

These plastic push-fit caps are designed for closing vials of injectable drugs. Flip-off snap caps are pre-assembled with an ISO 8362-2 compliant elastomer stopper to simplify the production process.
Supplier: RayDyLyo™ 255853002
This item is not returnable.
View return policy
Description
- Container Closure Integrity (CCI) validated for use with ISO 8362-1 glass and COP vials
- Delivered sterile (RTU) with a 2 year shelf life from time of manufacture
- Clean room manufactured with 100% in-process quality controls
- Suitable for aseptic small batch, clinical trial, and robotic fill and finish operations
- Snap caps do not require special crimping tools and affix with single vertical pressure
- Provides tamper-evident closing and eliminates crimping defects
- Compatible with gamma and steam terminal sterilization
- Available in nested, standard bag, and RTP-type bag configurations

Specifications
Red | |
Polypropylene | |
Nested | |
3600/Pk. | |
13 mm |
13 mm | |
Sterile | |
Autoclavable | |
ISO 8362-2 | |
Serum Vials |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RayDyLyo™ is a registered trademark of ARaymond.
Spot an opportunity for improvement?Share a Content Correction