Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAP23 Rabbit anti-Human, Clone: JA73-15, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP267157

Catalog No. NBP267157


Add to cart

Description

Description

SNAP23 Monoclonal antibody specifically detects SNAP23 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

SNAP23
JA73-15
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
HsT17016, SNAP-23, SNAP23A, SNAP23B, synaptosomal-associated protein 23, synaptosomal-associated protein, 23kD, synaptosomal-associated protein, 23kDa, Vesicle-membrane fusion protein SNAP-23
Rabbit
IgG
100μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin
Purified
SNAP23
Recombinant Protein
Protein A purified
Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Phosphatase
Primary
8773
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit