Learn More
SNAP25 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 149031AP
Catalog No. 50-172-9821
Add to cart
Description
Description
SNAP25 is a t-SNARE involved in the molecular regulation of neurotransmitter release. It may play a role in the synaptic function of specific neuronal systems. SNAP25 associates with proteins involved in vesicle docking and membrane fusion. It also regulates plasma membrane recycling through its interaction with CENPF. Mutations in the gene can results in myasthenic syndrome.
Specifications
Specifications
SNAP25
0.48 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P60879, P60880, P60881
RIC 4, RIC4, SEC9, SNAP, SNAP 25, SNAP25, SUP, Super protein
Rabbit
IgG
150 μL
-20°C
Primary
20614, 25012, 6616
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
SNAP25
Liquid
SNAP25
SNAP25 Fusion Protein Ag6695
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat