Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAP25, Rabbit anti-Human,Mouse,Rat,Porcine,Bovine,Feline,Guinea Pig,Hamster, Clone: SP12, Novus Biologicals™

Rabbit Monoclonal Antibody

$156.50 - $419.00

Specifications

Antigen SNAP25
Clone SP12
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Host Species Rabbit
Target Species Human, Mouse, Rat, Porcine, Bovine, Feline, Guinea Pig, Hamster
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB015045
View Documents
Novus Biologicals
NBP2810570.025MG
0.025 mg
Each for $156.50
Add to cart
 
NB015046
View Documents
Novus Biologicals
NBP2810570.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

SNAP25 Monoclonal antibody specifically detects SNAP25 in Human, Mouse, Rat, Porcine, Bovine, Feline, Guinea Pig, Hamster Samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence.
Specifications

Specifications

SNAP25
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Human, Mouse, Rat, Porcine, Bovine, Feline, Guinea Pig, Hamster
Primary
SP12
Rabbit
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit