Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNAP29 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | SNAP29 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17976920
![]() |
Novus Biologicals
NBP17976920UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179769
![]() |
Novus Biologicals
NBP179769 |
100 μL |
Each for $487.50
|
|
|||||
Description
SNAP29 Polyclonal specifically detects SNAP29 in Rat samples. It is validated for Western Blot.Specifications
SNAP29 | |
Polyclonal | |
Rabbit | |
NP_446262 | |
9342 | |
The immunogen for this antibody is Snap29. Peptide sequence NSIKSVFGGFINYFKSKPVEPPPEQNGSIVPQPSSRLKEAINTSKDQESK. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
CEDNIKSoluble 29 kDa NSF attachment protein, SNAP-29FLJ21051, synaptosomal-associated protein 29, synaptosomal-associated protein, 29kD, synaptosomal-associated protein, 29kDa, Vesicle-membrane fusion protein SNAP-29 | |
SNAP29 | |
IgG | |
29 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title