Learn More
SNAP29 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSS00034W
Catalog No. PIOSS00034W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water.
This gene, a member of the SNAP25 gene family, encodes a protein involved in multiple membrane trafficking steps. Two other members of this gene family, SNAP23 and SNAP25, encode proteins that bind a syntaxin protein and mediate synaptic vesicle membrane docking and fusion to the plasma membrane. The protein encoded by this gene binds tightly to multiple syntaxins and is localized to intracellular membrane structures rather than to the plasma membrane. While the protein is mostly membrane-bound, a significant fraction of it is found free in the cytoplasm. Use of multiple polyadenylation sites has been noted for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SNAP29
Polyclonal
Snap29
SNAP-29, Vesicle-membrane fusion protein, SNAP29, Soluble 29 kDa NSF attachment protein, Golgi SNARE of 32 kDa, Gs32
Rabbit
100 μL
Primary
Mouse, Rat
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
Q9ERB0, Q9Z2P6
SNAP29
A synthetic peptide from rat Synaptosomal-associated protein 29
RUO
116500, 67474
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations
Recommendations