Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNAP29 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP16918020UL
Description
SNAP29 Polyclonal specifically detects SNAP29 in Human samples. It is validated for Western Blot.Specifications
SNAP29 | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
O95721 | |
SNAP29 | |
Synthetic peptides corresponding to SNAP29 (synaptosomal-associated protein, 29kDa) The peptide sequence was selected from the middle region of SNAP29. Peptide sequence QPNNRLKEAISTSKEQEAKYQASHPNLRKLDDTDPVPRGAGSAMSTDAYP. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
CEDNIKSoluble 29 kDa NSF attachment protein, SNAP-29FLJ21051, synaptosomal-associated protein 29, synaptosomal-associated protein, 29kD, synaptosomal-associated protein, 29kDa, Vesicle-membrane fusion protein SNAP-29 | |
Rabbit | |
29 kDa | |
20 μL | |
Neuronal Cell Markers, Neurotransmission | |
9342 | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title