Learn More
Aviva Systems Biology Corp SNAPC2 Antibody - middle region (P100961_P050) | 100uL
Supplier:  Aviva Systems Biology Corp P100961_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNAPC2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SNAPC2. Alternate names: PTFdelta, SNAP45, PTFDELTAApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-62802