Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SND1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA540124
Description
Recommended positive controls: 293T and HepG2 cells. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (89%), Xenopus laevis (93%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SND1 Polyclonal antibody specifically detects SND1 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
SND1 | |
Polyclonal | |
Unconjugated | |
Snd1 | |
100 kDa coactivator; 4SNc-Tudor domain protein; 4SNc-Tudor domain protein long form; 4SNc-Tudor domain protein short form; AL033314; EBNA-2 co-activator (100kD); EBNA2 coactivator p100; p100; p100 co-activator; p100 co-activator variant 1; p100 EBNA2 co-activator; p100L; P100-like protein short variant; p100S; similar to Homo sapiens EBNA-2 co-activator (100kDa coactivator); p105 coactivator; SN4TDR; SND p102; SND1; snd1 protein; SND1-BRAF fusion; staphylococcal nuclease and tudor domain containing 1; staphylococcal nuclease domain containing 1; staphylococcal nuclease domain-containing protein 1; TDRD11; testis tissue sperm-binding protein Li 82P; tudor domain-containing protein 11; TudorSN; Tudor-SN; wu:fc27g10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27044 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
0.74 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q7KZF4 | |
Snd1 | |
Recombinant protein encompassing a sequence within the center region of human SND1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction