Learn More
Innovative Research Inc Sngl Hmn Ped Pla Age 11-14 1
Supplier: Innovative Research Inc IPLASPED11141
Single Donor Human Pediatric Plasma Ages 11-14 1 Sample - Single Donor Human Pediatric Plasma from Innovative Research is generally 0.5ml in sample size with each sample collected from a unique donor. Age groupings available include 0-3 years, 4-7 years, 8-10 years, 11-14 years and 15-17 years, or you may elect unspecified (any age up to 18 years). Anticoagulant used is K2 EDTA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.