Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNIP1 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer: Abnova Corporation H00079753T01
Description
Western Blotting
Specifications
Western Blot | |
SNIP1 293T Cell Transient Overexpression Lysate(Denatured) | |
43.67 | |
pCMV-SNIP1 full-length | |
100 μL | |
Human |
293T | |
BC027040 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-SNIP1 antibody (H00079753-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title