Learn More
Aviva Systems Biology Corp SNRK Antibody - middle region (ARP56306_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56306_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SNRK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SNRK. Alternate names: DKFZp779A1866, FLJ20224, HSNFRK, KIAA0096ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61803