Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNRPB Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 168071AP

Catalog No. 50-173-1189


Add to cart

Description

Description

The protein encoded by this gene is one of several nuclear proteins that are found in common among U1, U2, U4/U6, and U5 small ribonucleoprotein particles (snRNPs). These snRNPs are involved in pre-mRNA splicing, and the encoded protein may also play a role in pre-mRNA splicing or snRNP structure. Autoantibodies from patients with systemic lupus erythematosus frequently recognize epitopes on the encoded protein. Two transcript variants encoding different isoforms (B and B ft.) have been found for this gene.
Specifications

Specifications

SNRPB
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P14678
COD, Sm B/B, Sm protein B/B, SmB/B, SmB/SmB′, snRNP B, SNRPB, SNRPB1
Rabbit
IgG
150 μL
-20°C
Primary
6628
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SNRPB
Liquid
SNRPB
SNRPB Fusion Protein Ag10298
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit